Anti-NGFRAP1 Rabbit Polyclonal Antibody (Cy5.5®)
Fournisseur: Bioss
BS-7671R-CY5.5
BOSSBS-7671R-CY5.5EA 900 EUR InStock
BOSSBS-7671R-CY5.5
Anti-NGFRAP1 Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
May be a signaling adapter molecule involved in p75NTR-mediated apoptosis induced by NGF. Plays a role in zinc-triggered neuronal death (By similarity). May play an important role in the pathogenesis of neurogenetic diseases.Tissue specificity: Found in ovarian granulosa cells, testis, prostate and seminal vesicle tissue. High levels also detected in liver.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NGFRAP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité