Anti-CDC123 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-7781R
BOSSBS-7781REA 725 EUR InStock
BOSSBS-7781R
Anti-CDC123 Rabbit Polyclonal Antibody
Anticorps
Required for S phase entry of the cell cycle.The eukaryotic cell division cycle consists of a number of gene-controlled sequences that involve cyclin dependent kinases (Cdks) and cell division control (Cdc) proteins. Cdc123 (Cell division cycle protein 123), also known as D123, is a 336 amino acid cytoplasmic protein that is involved in cell cycle control. Widely expressed with high expression in thymus, spleen, ovary, testis, small intestine and leukocytes, Cdc123 functions to destabilize Chfr (checkpoint with forkhead and ring finger domain) proteins which, when accumulated, block the G toMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité