Anti-CLASP2 Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-7793R-CY3
BOSSBS-7793R-CY3EA 900 EUR InStock
BOSSBS-7793R-CY3
Anti-CLASP2 Rabbit Polyclonal Antibody (Cy3®)
Anticorps
CLASP2 is a microtubule plus end tracking protein that promotes the stabilization of dynamic microtubules and is required for the polarization of the cytoplasmic microtubule arrays in migrating cells towards the leading edge of the cell. CLASP2 may act at the cell cortex to enhance the frequency of rescue of depolymerizing microtubules by attaching their plus ends to cortical platforms composed of ERC1 and PHLDB2. This cortical microtubule stabilizing activity is regulated at least in part by phosphatidylinositol 3 kinase signaling. CLASP2 also performs a similar stabilizing function at theMore Product Information
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-7793R-CY3
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité