Anti-CA7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-7984R-A647
BOSSBS-7984R-A647EA 893 EUR InStock
BOSSBS-7984R-A647
Anti-CA7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
Carbonic anhydrases are a large family of zincmetalloenzymes that catalyze the reversible hydration of carbondioxide. They participate in a variety of biological processes,including respiration, calcification, acid-base balance, boneresorption, and the formation of aqueous humor, cerebrospinalfluid, saliva, and gastric acid. They show extensive diversity intissue distribution and in their subcellular localization. Thecytosolic protein encoded by this gene is predominantly expressedin the salivary glands. Alternative splicing in the coding regionresults in multiple transcript variantsMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité