Anti-PLEKHM3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: Bioss
BS-8064R-HRP
BOSSBS-8064R-HRPEA 900 EUR InStock
BOSSBS-8064R-HRP
Anti-PLEKHM3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
PLEKHM3 (pleckstrin homology domain containing, family M, member 3), also known as DAPR or PLEKHM1L (pleckstrin homology domain containing, family M, member 1-like), is a 761 amino acid phosphoprotein that contains two pleckstrin homology (PH) domains and one phorbol-ester/DAG-type zinc finger. Conserved in chimpanzee, dog, cow, mouse, rat, chicken and zebrafish, PLEKHM3 exists as three alternatively spliced isoforms that participate in metal ion binding. The gene that encodes PLEKHM3 maps to human chromosome 2q33.3. As the second largest human chromosome, chromosome 2 makes up approximatelyMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité