Anti-CCDC58 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-8080R-A350
BOSSBS-8080R-A350EA 893 EUR InStock
BOSSBS-8080R-A350
Anti-CCDC58 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
The coiled-coil domain is a structural motif found in proteins that are involved in a diverse array of biological functions such as the regulation of gene expression, cell division, membrane fusion and drug extrusion and delivery. CCDC58 (coiled-coil domain containing 58) is a 144 amino acid protein that is encoded by a gene that maps to human chromosome 3q21.1. Chromosome 3 is made up of about 214 million bases encoding over 1,100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3More Product Information
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-8080R-A350
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité