Anti-DDRGK1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: Bioss
BS-8252R-FITC
BOSSBS-8252R-FITCEA 900 EUR InStock
BOSSBS-8252R-FITC
Anti-DDRGK1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
DDRGK1 (DDRGK domain-containing protein 1), also known as C20orf116, is a 314 amino acid secreted protein. DDRGK1 contains one PCI domain and is expressed as two isoforms produced by alternative splicing. The gene that encodes DDRGK1 maps to human chromosome 20, which represents about 2% of human DNA and consists of approximately 63 million bases and 600 genes. Chromosome 20 contains a region with numerous genes expressed in the epididymis, which are thought to be important for seminal production. The PRNP gene encoding the prion protein associated with spongiform encephalopathies, likeMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité