Anti-GPRIN2 Rabbit Polyclonal Antibody (Cy5.5®)
Fournisseur: Bioss
BS-8276R-CY5.5
BOSSBS-8276R-CY5.5EA 900 EUR InStock
BOSSBS-8276R-CY5.5
Anti-GPRIN2 Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
G protein-coupled receptors (GPCRs) represent a large superfamily of cell-surface receptors that are involved in a multitude of physiological processes such as perception of sensory information, modulation of synaptic transmission, hormone release/actions, regulation of cell contraction/migration and cell growth/differentiation. GPCRs interact with G proteins (heterotrimeric GTPases) to synthesize intracellular second messengers, such as diacylglycerol, cyclic AMP, inositol phosphates and calcium ions. Their diverse biological functions range from vision and olfaction to neuronal andMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité