Anti-DPYSL3 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-8285R-CY7
BOSSBS-8285R-CY7EA 893 EUR InStock
BOSSBS-8285R-CY7
Anti-DPYSL3 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
The collapsin response mediator protein (CRMP) family of five cytosolic phosphoproteins are highly expressed throughout brain development. The functions of CRMPs encompass signal transduction in developmental guidance cues as well as multiple cellular and molecular events involved in apoptosis/proliferation, cell migration, and differentiation. In the adult brain, the expression of CRMPs is dramatically downregulated. However, CRMPs remain expressed in structures that retain their capacity for differentiation and plasticity. The expression of CRMPs is altered in neurodegenerative diseases,More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité