Anti-PANK3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-8339R-A647
BOSSBS-8339R-A647EA 893 EUR InStock
BOSSBS-8339R-A647
Anti-PANK3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
The pantothenate kinase (PANK) family of proteins catalyzes the first step in coenzyme A (CoA) biosynthesis. Coenzyme A is an important coenzyme involved in the synthesis and oxidation of fatty acids, as well as the oxidation of pyruvate in the citric acid (Krebs) cycle. Pantothenate kinase 3 (PANK3) is a 370 amino acid member of the pantothenate kinase family that plays a role in the physiological regulation of the intracellular CoA concentration. Localized to the cytoplasm, PANK3 is regulated by feedback inhibition by CoA and its thioesters. PANK3 transfers a phosphate from ATP toMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité