Anti-Ube2N Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-8351R-A680
BOSSBS-8351R-A680EA 893 EUR InStock
BOSSBS-8351R-A680
Anti-Ube2N Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). UBC13, also known as UBE2N or BLU, is a 152 amino acid member of the E2 ubiquitin-conjugating enzyme family. Existing as a heterodimer with Mms2 (also known as UBE2V2), UBC13 catalyzes the ATP-dependent synthesis of non-canonical polyubiquitin chains, a process that does not lead to proteasomal degradation. Additionally, UBC13 mediates the transcription of several target genes and is thought to play a role in cell cycle progression, cellular differentiation and DNA repair mechanisms that ensure cell survival after DNA damage.
Type: Primary
Antigen: Ube2N
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 56-152/152
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Others
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité