Anti-FBXO15 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Fournisseur: Bioss
BS-8396R-A555
BOSSBS-8396R-A555EA 893 EUR InStock
BOSSBS-8396R-A555
Anti-FBXO15 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
FBXO15, also known as FBX15, is a 434 amino acid protein that contains one C-terminal F-box domain and belongs to the F-box family of proteins. F-box proteins are critical components of the SCF (Skp1-CUL-1-F-box protein) type E3 ubiquitin ligase complex and are involved in substrate recognition and recruitment for ubiquitination. They are members of a larger family of proteins that are involved in the regulation of a wide variety of cellular processes (including the cell cycle, immune response, signaling cascades and developmental processes) through the targeting of proteins, such as cyclins,More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité