Anti-CENPK Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Fournisseur: Bioss
BS-8459R-A555
BOSSBS-8459R-A555EA 893 EUR InStock
BOSSBS-8459R-A555
Anti-CENPK Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
FKSG14, also known as CENPK (centromere protein K) is a component of the CENPA-CAD (nucleosome distal) complex. It may be involved in incorporation of CENPA into centromeres and is required for proper kinetochore function, mitotic progression and chromosome segregation. May be involved in incorporation of newly synthesized CENPA into centromeres via its interaction with the CENPA-NAC complex. Acts in coordination with CASC5/KNL1 to recruit the NDC80 complex to the outer kinetochore. FKSG14 constitutively localized to centromeres throughout the cell cycle. There are 3 isoforms produced byMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité