Anti-PMS1 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-8579R-CY7
BOSSBS-8579R-CY7EA 900 EUR InStock
BOSSBS-8579R-CY7
Anti-PMS1 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
The finding that mutations in DNA mismatch repair genes are associated with hereditary nonpolyposis colorectal cancer (HNPCC) has resulted in considerable interest in the understanding of the mechanism of DNA mismatch repair. Initially, inherited mutations in the MSH2 and MLH1 homologs of the bacterial DNA mismatch repair genes MutS and MutL were demonstrated at high frequency in HNPCC and were shown to be associated with microsatellite instability. The demonstration that 10 to 45% of pancreatic, gastric, breast, ovarian and small cell lung cancers also display microsatellite instability hasMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité