Anti-G1switch 2 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-8656R
BOSSBS-8656REA 725 EUR InStock
BOSSBS-8656R
Anti-G1switch 2 Rabbit Polyclonal Antibody
Anticorps
G0S2 is a 103 amino acid novel target of peroxisome proliferator-activated receptors (PPARs) and regulator of latent HIV. G0S2 may be involved in adipocyte differentiation and its expression is essential for committing cells to enter the G1 phase of the cell cycle. G0S2 contains a CpG-rich island and multiple sites for potential phosphorylation by casein kinase II and protein kinase C. The gene encoding G0S2 maps to human chromosome 1, which is the largest human chromosome. Chromosome 1 spans about 260 million base pairs and makes up 8% of the human genome. There are about 3,000 genes onMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité