Anti-MMS19 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: Bioss
BS-8671R-FITC
BOSSBS-8671R-FITCEA 893 EUR InStock
BOSSBS-8671R-FITC
Anti-MMS19 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
Key component of the cytosolic iron-sulfur protein assembly (CIA) complex, a multiprotein complex that mediates the incorporation of iron-sulfur cluster into apoproteins specifically involved in DNA metabolism and genomic integrity. In the CIA complex, MMS19 acts as an adapter between early-acting CIA components and a subset of cellular target iron-sulfur proteins such as ERCC2/XPD, FANCJ and RTEL1, thereby playing a key role in nucleotide excision repair (NER) and RNA polymerase II (POL II) transcription. As part of the mitotic spindle-associated MMXD complex, plays a role in chromosomeMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité