Anti-ORC1 Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-8998R-CY5
BOSSBS-8998R-CY5EA 900 EUR InStock
BOSSBS-8998R-CY5
Anti-ORC1 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
The initiation of DNA replication is a multi-step process that depends on the formation of pre-replication complexes, which trigger initiation (1). Among the proteins required for establishing these complexes are the origin recognition complex (ORC) proteins (1). ORC proteins bind specifically to origins of replication where they serve as scaffold for the assembly of additional initiation factors (1). Human ORC subunits 1-6 are expressed in the nucleus of proliferating cells and tissues, such as the testis (2). ORC1 and ORC2 are both expressed at equivalent concentrations throughout the cellMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité