Anti-RCBTB1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-9007R-A350
BOSSBS-9007R-A350EA 893 EUR InStock
BOSSBS-9007R-A350
Anti-RCBTB1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
The BTB domain, also known as the POZ (poxvirus and zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. RCBTB1 (regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 1), also known as GLP, CLLD7, CLLL7 or E4.5, is a 531 amino acid protein that localizes to the nucleus and contains two BTB (POZ) domains and six RCC1 repeats. ExpressedMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité