Anti-DIS3L Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Fournisseur: Bioss
BS-9052R-A555
BOSSBS-9052R-A555EA 893 EUR InStock
BOSSBS-9052R-A555
Anti-DIS3L Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
The exosome is a multi-protein complex composed of several highly conserved subunits, some of which are 3’ to 5’ exoribonucleases. The complex is involved in a variety of cellular processes and is responsible for degrading unstable mRNAs that contain AU-rich elements (AREs) in their untranslated 3’ regions. DIS3, also known as RRP44, is a 958 amino acid protein that localizes to both the cytoplasm and the nucleus and contains one PINc domain. Widely expressed with highest expression in testis, DIS3 functions as a component of the exosome exoribonuclease complex and is required forMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité