Anti-RNF16 Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-9161R-CY5
BOSSBS-9161R-CY5EA 900 EUR InStock
BOSSBS-9161R-CY5
Anti-RNF16 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
The tripartite motif (TRIM) family of proteins are characterized by a conserved TRIM domain that includes a coiled-coil region, a B-box type zinc finger, one RING finger and three zinc-binding domains. TRIM17 (tripartite motif-containing 17), also known as RBCC, terf or RNF16, is a 477 amino acid protein that contains one RING-type zinc finger, one SPRY domain and one B box-type zinc finger. Expressed nearly exclusively in testis, TRIM17 belongs to the TRIM family and, based on its functional domains, may play a role in transcriptional regulation events. The gene encoding TRIM17 maps toMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité