Anti-RNF113B Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-9256R-A350
BOSSBS-9256R-A350EA 893 EUR InStock
BOSSBS-9256R-A350
Anti-RNF113B Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. RNF113B (ring finger protein 113B), also referred to as zinc finger protein 183-like 1, RNF161, MGC26599, bA10G5.1 or ZNF183L1, is a 322 amino acid protein containing one C3H1-type zinc finger and one RING-type zinc finger. The gene encoding RNF113B maps to human chromosome 13, which houses over 400More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité