Anti-KATNA1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: Bioss
BS-9308R-FITC
BOSSBS-9308R-FITCEA 893 EUR InStock
BOSSBS-9308R-FITC
Anti-KATNA1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
Microtubules are polymers of alpha and beta subunits that form the mitotic spindle and assist in the organization of membranous organelles during interphase. Katanin p60 A1, also known as KATNA1, is a 491 amino acid protein that belongs to the AAA ATPase family and is involved in microtubule regulation. Localized to the cytoplasm and to the centrosome, Katanin p60 A1 functions to sever and disassemble microtubules in an ATP-dependent manner, thus promoting the rapid reorganization of cellular microtubule arrays and playing an important role in microtubule release from the centrosome afterMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité