Anti-RANBP2 Rabbit Polyclonal Antibody (Cy5.5®)
Fournisseur: Bioss
BS-9323R-CY5.5
BOSSBS-9323R-CY5.5EA 900 EUR InStock
BOSSBS-9323R-CY5.5
Anti-RANBP2 Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
The nuclear pore complex protein, Ran-binding protein 2 (Ran BP-2 or Nup358), contains four Ran-binding domains. Ran BP-2 is a large scaffold cyclophilin-related protein expressed in photoreceptor cells. Ran BP-2 localization at cytoplasmic fibrils emanating from the nuclear pore complex and interaction with the Ran-GTPase support its role in nucleocytoplasmic transport processes. In humans, the Ran BP-2 gene is partially duplicated in a gene cluster and lies in a hot spot for recombination on chromosome 2q. This genetic heterogeneity renders further significance of this genomic region inMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité