Anti-MARCH3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: Bioss
BS-9337R-HRP
BOSSBS-9337R-HRPEA 900 EUR InStock
BOSSBS-9337R-HRP
Anti-MARCH3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). MARCH3 (membrane-associated ring finger (C3HC4) 3), also known as RNF173, is a 253 amino acid multi-pass membrane protein that localizes to cytoplasmic vesicles and early endosomes and contains one RING-CH-type zinc finger. Involved in the pathway of proteinMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité