Anti-PSMD7 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-9366R
BOSSBS-9366REA 725 EUR InStock
BOSSBS-9366R
Anti-PSMD7 Rabbit Polyclonal Antibody
Anticorps
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in aMore Product Information
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
100 μg 100 µl BOSSBS-9366R
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité