Anti-PSMD6 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-9367R-CY7
BOSSBS-9367R-CY7EA 900 EUR InStock
BOSSBS-9367R-CY7
Anti-PSMD6 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
In eukaryotic cells, selective breakdown of cellular proteins is ensured by two distinct pathways, ubiquitination and degradation by the 26S proteasome. At specific stages of development, embryo- and tissue-specific components of the 26S proteasome are formed by developmentally regulated alternative splicing, including Rpn10a through Rpn10e (also designated pUb-R2 through pUb-R5). The pUb-R2 subunit, originally identified as S5a, is ubiquitously expressed and may perform proteolysis constitutively in a wide variety of cells. p44S10 is a highly conserved proteasome regulatory subunit that isMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité