Anti-CHAF1A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-9392R-A647
BOSSBS-9392R-A647EA 893 EUR InStock
BOSSBS-9392R-A647
Anti-CHAF1A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
Core component of the CAF-1 complex, a complex thought to mediate chromatin assembly in DNA replication and DNA repair. Assembles histone octamers onto replicating DNA in vitro. CAF-1 performs the first step of the nucleosome assembly process, bringing newly synthesized histones H3 and H4 to replicating DNA; histones H2A/H2B can bind to this chromatin precursor subsequent to DNA replication to complete the histone octamer. CHAF1A binds to histones H3 and H4. It may play a role in heterochromatin maintenance in proliferating cells by bringing newly synthesized cbx proteins to heterochromaticMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité