Anti-TSPAN3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Fournisseur: Bioss
BS-9412R-A555
BOSSBS-9412R-A555EA 893 EUR InStock
BOSSBS-9412R-A555
Anti-TSPAN3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
TSPAN3 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene. Two alternative transcripts encoding different isoforms have been described.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: TSPAN3More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité