Anti-TSPAN9 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-9448R-CY7
BOSSBS-9448R-CY7EA 893 EUR InStock
BOSSBS-9448R-CY7
Anti-TSPAN9 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
The tetraspanin family is a group of cell surface proteins that regulate cell development, activation, growth and motility. Each member contains four hydrophobic domains and participates in the mediation of signal transduction. NET-5, also known as TSPAN9 (tetraspanin 9), is a 239 amino acid multi-pass membrane protein that belongs to the tetraspanin (TM4SF) family. NET-5 forms a complex with GPVI in the tetraspanin microdomains on the platelet surface, and is encoded by a gene that maps to human chromosome 12p13.33. Chromosome 12 encodes over 1,100 genes and comprises approximately 4.5% ofMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité