Anti-SLC27A6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-9454R-A680
BOSSBS-9454R-A680EA 893 EUR InStock
BOSSBS-9454R-A680
Anti-SLC27A6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Acyl-coenzyme A synthetases (ACSs) are a large family of related enzymes known to catalyse the fundamental initial reaction in fatty acid metabolism. The ACS family is roughly characterised based on fatty acid chain length preference amongst different members. The nomenclature in the ACS family reflects this relationship and includes short-chain ACS (ACSS), medium-chain ACS (ACSM), long-chain ACS (ACSL) and very long-chain ACS (ACSVL). ACSVL family members are capable of activating both long (LCFAs) and very long-chain fatty acids (VLCFAs). There are six members of the human ACSVL subfamily,More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité