Anti-C21orf55 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-9980R-A750
BOSSBS-9980R-A750EA 893 EUR InStock
BOSSBS-9980R-A750
Anti-C21orf55 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
The smallest of the human chromosomes, 21 makes up about 1.5% of the human genome. Chromosome 21 contains nearly 300 genes and 47 million base pairs. Down syndrome, also known as trisomy 21, is the disease most commonly associated with chromosome 21. Alzheimer's disease, Jervell and Lange-Nielsen syndrome and amyotrophic lateral sclerosis are also associated with chromosome 21. Translocations are found to occur between chromosome 21 and 8, and chromosome 21 and 12, in certain leukaemias. DnaJC28 (DnaJ homolog subfamily C member 28), also known as C21orf55, C21orf78 or MGC27620 is a 388 aminoMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité