Introcan Safety® IV Catheter
Fournisseur: B. Braun
4252500-01
BRAU4252500-01EA 653 EUR InStock
BRAU4252500-01
Introcan Safety® IV Catheter
Cathéters
Passive safety technology is incorporated into the Introcan Safety® IV catheter via an integrated fully automatic safety shield which protects the needle tip to prevent needlestick injuries.
  • Easy puncture at a wide range of angles
  • Minimum effort of catheter insertion
  • Simplicity - looks and feels like a standard cannula
  • Self-activating safety shield - covers needle tip automatically after use
It was confirmed that passive safety engineered devices create significantly better protection for healthcare workers than those that require the user to activate the safety feature.
Passive safety devices were associated with the lowest needlestick injury rate and are most effective for needlestick injury prevention.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité