Labels for Thermal Transfer Printers, Type B-413, MetaLabel
Fournisseur: Brady
104170
BRDY104170EA 539.19 EUR InStock
BRDY104170
Labels for Thermal Transfer Printers, Type B-413, MetaLabel
Étiquettes
These metallic, non metal labels exceed expectations in most industrial environments. They are designed to work with the easy-to-use software and the full line of thermal transfer printers. These materials will allow you to create the sharp, professional rating plates and other labels you need.
  • Print faster, easier, more efficient
  • Easy to wipe clean label front
These labels coupled with Brady's IP™ Thermal Transfer Printing System, are the only metallic product available for hassle-free printing. Supplied with industrial-strength adhesive, as well as Mondo Bondo™ adhesive, these labels can be trusted in dirty, oily environments.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité