Brady® R6400 Printer Ribbon for BBP11 and BBP12 Label Printers
Fournisseur: Brady
112648
BRDY112648EA 89.5 EUR InStock
BRDY112648
Brady® R6400 Printer Ribbon for BBP11 and BBP12 Label Printers
Rubans pour imprimante
This ink-side-out ribbon features an open 12,7 mm core and is to be used for the Brady BBP11 label printer and BBP12 label printer.
  • One-colour ribbon for printing a single colour onto a label
  • R6400 series resin formulation offers superior chemical and abrasion resistance when printed on the appropriate label material from Brady®
  • Combine brady label materials with their recommended ribbon series for superior legend performance
Recommended for use with materials designed to resist chemicals, such as B-492 frozen surface polyester and B-481 chemical resistant polyester.
Application: Barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, inventory and inspection labelling, laboratory labelling, wire and cable labelling
Resistance properties: Abrasion-resistant, chemical-resistant, halogen free
Core diameter: 12,7 mm
Certifications: Compliant with Directive 2011/65/EU and 2015/863.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité