Sign and label printer with Wifi - AZERTY, BradyPrinter S3100
Fournisseur: Brady
149122
BRDY149122EA 1762.8 EUR InStock
BRDY149122
Sign and label printer with Wifi - AZERTY, BradyPrinter S3100
Imprimantes Label Printers
This S3100 printer is extremely easy to use and features auto-calibration and automatic label material setup
This will enable you to just walk up to the printer and start printing the signs and labels you need. Switching to another label material is easy and extremely fast thanks to the printer's 'drop-in' label materials and matching ink ribbons that can be switched out in less than 20 seconds. An intuitive user interface with touch screen offers a wide variety of sign & label design options, even when the printer is used in standalone mode.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité