Workstation safety and facility identification suite on CD
Fournisseur: Brady
149200
BRDY149200EA 299.99 EUR InStock
BRDY149200
Workstation safety and facility identification suite on CD
Logiciels Logiciels de laboratoire
Keep your facility safe and compliant with the safety and facility identification suite. This group of apps makes it easy to create custom signs, labels and pipemarkers.
  • Hardware: 1 gigahertz (GHZ) processor or faster
  • RAM: 2GB
  • Screen resolution: 1024×768 or higher (recommended), 800×600 high colour (minimum)
Design and print labels for gauges, gradient scales and more in support of visual workplace initiatives - compatible with BBP37 only.
This group of apps makes it easy to create custom signs, labels and custom designer, express signs, arc flash, arrow tap, pipemarker, lean visuals, template editer.
Applicable with Windows® 10;Windows® 7 (32- or 64-bit);Windows® 7 Enterprise;Windows® 7 Professional;Windows® 7 SPI;Windows® 7 Ultimate;Windows® 8 (32- or 64-bit);Windows® 8 Professional (32 or 64 bit);Windows® 8.1 (32- or 64-bit);Windows® 8.1 Enterprise;Windows® 8.1 Professional.
Languages available: Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English (US), Estonian, Finnish, French (France), German, Hungarian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (Portugal), Romanian, Russian, Slovak, Slovenian, Spanish (Mexico), Spanish (Spain), Swedish, Turkish, Vietnamese.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité