Brady® SPC® Absorbents HAZWIK Quick Response Kit, Chemical
Fournisseur: Brady
152186
BRDY152186EA 220.44 EUR InStock
BRDY152186
Brady® SPC® Absorbents HAZWIK Quick Response Kit, Chemical
Matériaux absorbants
Get fast and effective spill cleanup with a HAZWIK® quick response kit. It contains everything you need for emergency spills in a convenient, grab-and-go-style bag.
  • Compact, single-use kit that is ideal for use on small spills in the laboratory
  • Quickly cleans up hazardous chemical spills
  • Visual instructions on the package
  • Contains PPE, tools, absorbent pads, acid and base neutralisers, wipes and disposable materials
Information de livraison: Includes 0,13 L acid neutraliser (1), 0,13 L base neutraliser (1), cable tie (1), disposal bag (1), pair of nitrile gloves (1), scoop and scraper (1), surface sanitising wipe (1), 190,5×254 mm pads (6).
Conditionnement: Packed as five individual kits per case.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité