Aggressive Adhesive Multi-Purpose Polyester Labels with Ribbon for M510, BMP51, BMP53 and M511 Printers
Fournisseur: Brady
170859
BRDY170859EA 76.07 EUR InStock
BRDY170859
Aggressive Adhesive Multi-Purpose Polyester Labels with Ribbon for M510, BMP51, BMP53 and M511 Printers
Étiquettes
Easily label components with the thermal transfer, glossy permanent polyester label material (B-422). Print almost anywhere with a durable Brady handheld label printer.
  • Versatile - Designed for a wide range of applications such as labeling equipment, circuit boards, panel components and barcodes
  • Tough - Constructed to withstand abrasions, extreme temperatures and solvents such as alcohol, oil and water
  • Adhesive - Made with a permanent acrylic adhesive that sticks to rough, smooth and powder-coated flat surfaces
Ideal for asset tracking, barcode labelling, component and equipment labelling, data and telecommunications labelling, general identification, inventory and inspection labelling applications.
Gloss finish, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, weather-resistant, outdoor use B-422 labels are compatible with Brady printer models M510, BMP51, BMP53, and M511.
Certifications: UL Recognized to UL969, CSA to C22.2 No.0.15-95, RoHS Compliant to 2011/65/EU (Pass 2015/863).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité