T-Flag or P-Flag Matte Polypropylene Label with Ribbon for M510, BMP51, BMP53 and M511 Printers
Fournisseur: Brady
170860
BRDY170860EA 52.61 EUR InStock
BRDY170860
T-Flag or P-Flag Matte Polypropylene Label with Ribbon for M510, BMP51, BMP53 and M511 Printers
Étiquettes
White on black B-425 thermal transfer printable polypropylene is ideal for labelling applications requiring excellent solvent resistance and print performance.
  • No visible affect to label when exposed to most common chemicals, oils or water
  • Service temperature of −70 to 80 °C (−94 to 176 °F)
  • Matte finish
Ideal for asset tracking, barcode labelling, component and equipment labelling, data and telecommunications labelling, general identification, inventory and inspection labelling, laboratory labelling, and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material.
Matte finish, abrasion-resistant, chemical-resistant, humidity-resistant, indoor use B-425 labels are compatible with Brady printer models M510, BMP51, BMP53, and M511.
Certifications: UL recognised. CSA accepted. Halogen-free (DIN VDE 0472/part 815).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité