Aggressive Adhesive Multi-Purpose Nylon Labels B-499 with Ribbon for M510, BMP51, BMP53, M511 Printers
Fournisseur: Brady
170928
BRDY170928EA 0 EUR InStock
BRDY170928
Aggressive Adhesive Multi-Purpose Nylon Labels B-499 with Ribbon for M510, BMP51, BMP53, M511 Printers
Étiquettes
Aggressive adhesive multi-purpose nylon labels is a tough and versatile label.
  • Versatile - Ideal for labeling tightly-curved surfaces such as wires and cables, as well as general-purpose lab, equipment panels, faceplates and terminal blocks
  • Durable - Designed to withstand extreme temperatures, chemicals, greases, oils, cleaners and detergents
  • Adhesive - Permanent adhesive labels that stick to smooth, rough and powder-coated surfaces alike
Application: Component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, wire and cable labelling.
Matte finish, abrasion-resistant, chemical-resistant, humidity-resistant, UV-resistant, weather-resistant, indoor use B-499 labels are compatible with Brady printer models M510, BMP51, BMP53, and M511.
Certifications: UL Recognized to UL969, CSA to C22.2 No. 0.15-95, RoHS compliant to 2011/65/EU (Pass 2015/863).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité