Label Printer EMEA, M511
Fournisseur: Brady
170970
BRDY170970EA 365 EUR InStock
BRDY170970
Label Printer EMEA, M511
Imprimantes Label Printers
Work smarter. Label larger with the M511. Print bigger, clearer messages. Go edge to edge. Design and print on the fly, or a PC. Plus, share files, import data and connect up to 5 users simultaneously.
  • Monocolour
  • Ideal for larger applications - print up to 38,10 mm labels
  • Less waste - go edge to edge with text, graphics and barcodes
  • Longer battery life with an internal Li-ION battery - 8+ hours of work; 1000 labels on a single charge
  • More durable - resists 1,8 m drops, shocks, 115 kg crushes and blowing sand and dust (MIL-STD-810G)
  • Prints anywhere - Bluetooth 5,0 (20 m range); connects up to 5 users simultaneously
  • Uses label creation software - Express Labels app via Bluetooth on a phone/tablet; Brady Workstation on a PC
  • Saves time; saves space - features an auto label cutter and prints upright or on its side
Applications include asset tracking, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, facility identification, general identification, inventory and inspection labelling, laboratory labelling, lean and 5S labelling, safety identification, warehouse marking, wire and cable labelling.
Certifications: RoHS 2011/65/EU, 2015/863, UKCA, cULus (US/Canada), FCC.
Information de livraison: This printer ships in a carton box with M5C-1500-595-WT-BK label cartridge, AC adapter with EU and UK plugs, Quick Start guide and a Brady Workstation Basic design suite.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité