High Tensile Polypropylene Inserts/Tags for M710 and BMP71
Fournisseur: Brady
173322
BRDY173322EA 84.45 EUR InStock
BRDY173322
High Tensile Polypropylene Inserts/Tags for M710 and BMP71
Étiquettes
Polypropylene tag stock (B-412) is ideal for wire and cable identification and product inventory identification, where legibility and tensile strength are needed.
  • Non-adhesive material is thermal transfer printable
  • Good abrasion and chemical resistance
  • Service temperature of −40 to +38 °C (−40 to 100 °F)
  • Matte finish
Applications include component and equipment labelling, data and telecommunications labelling, electrical labelling, and wire and cable labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6200.
Conditionnement: 750 labels per box.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité