Multipurpose Tear-Resistant Tags for M610, M611, BMP61, M710 and BMP71
Fournisseur: Brady
173444
BRDY173444EA 350.58 EUR InStock
BRDY173444
Multipurpose Tear-Resistant Tags for M610, M611, BMP61, M710 and BMP71
Étiquettes
Durable non-adhesive polyethylene (B-109) is a multipurpose, matte finish tag material that can be used for a variety of tag applications.
  • Extremely tear resistant and able to withstand cold temperatures
  • Ideal material for both thermal transfer and dot matrix printing methods
  • Service temperature of −40 to +49 °C (−40 to +120 °F)
  • Matte finish
Applications include data and telecommunications labelling, wire and cable labelling, inventory and inspection labelling, lockout tagout, and safety identification.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R4300 for the M710/BMP71 label printers.
Certifications: RoHS compliant.
Conditionnement: 1000 labels per box.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité