Fire Retardant Oval Tags for M710 and BMP71
Fournisseur: Brady
174538
BRDY174538EA 130.56 EUR InStock
BRDY174538
Fire Retardant Oval Tags for M710 and BMP71
Étiquettes
Rigid polyethylene tags (B-145) are printable, non-adhesive, tear-resistant, flame retardent tags used for a wide array of identification applications.
  • Constructed from a 9,5 mil, cross laminated polyethylene film and a thermal transfer printable topcoat
  • Suitable for thermal transfer printing, then folded over and held together in a two-ply format with a pressure sensitive adhesive
  • Operating temperature of −40 to +100 °C (−40 to +212 °F)
  • Matte finish
Applications include asset tracking, component and equipment labelling, data and telecommunications labelling, facility identification, general identification, inventory and inspection labelling, safety identification, and wire and cable labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6200.
Certifications: RoHS compliant.
Conditionnement: 100 labels per box.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité