Rechargeable NiMH Battery for M510/M610/M710 Printers
Fournisseur: Brady
175674
BRDY175674EA 97.71 EUR InStock
BRDY175674
Rechargeable NiMH Battery for M510/M610/M710 Printers
Piles
Work longer with this NiMH M510, M610 and M710 battery pack. It enables cordless printer operation so you can print labels where (and when) you need them.
  • Removable, rechargeable Nickel-metal hydride battery pack
  • Battery can be charged only while installed in an M510, M610 or M710 printer
  • Prints up to 1,700 labels per charge
Work longer with this NiMH M510, M610 and M710 Battery Pack. It enables cordless printer operation so you can print labels where (and when) you need them. Removable, rechargeable Nickel-metal Hydride Battery Pack. Battery can be charged only while installed in an M510, M610 or M710 printer. Prints up to 1,700 labels per charge.
Informations de commande: Order reference M-NIMH-BATTERY.
Information de livraison: NiMH battery pack.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité