Printable Polyester Safety Tags for B30 Printers
Fournisseur: Brady
176270
BRDY176270EA 266.34 EUR InStock
BRDY176270
Printable Polyester Safety Tags for B30 Printers
Étiquettes
Rigid, high-quality polyester tags are perfect for marking on-the-spot warnings of dangerous conditions.
  • Print on one side, write anywhere, to easily customise messaging
  • Exceeds OSHA's 50 lb. pull force requirement for tagout devices
  • Textured write-on surface allows for authorised signatures, dates, and other special instructions
  • Superior durability over paper cardstock and withstands moisture, grease, and dirt
  • Rectangular label with oval hole
  • Die-cut
Create professional, impactful safety tags by adding specific hazard information to these print-on, write-on tags. These tags comply with OSHA 1910.145(f) when printed with required signal word and major message.
Labelling tags are compatible with BBP30, BBP31, BBP33, BBP35, BBP37, i3300, S3000, S3100, S3700, and i5300 series printers. A ribbon is required to print this material. Use part number B30-R10000.
Certifications: Conforms to OSHA Specification 1910.145.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité