Brady® P-Flag Matte Polypropylene Labels for M410, M510, and M511 Printers
Fournisseur: Brady
177005
BRDY177005EA 47.31 EUR InStock
BRDY177005
Brady® P-Flag Matte Polypropylene Labels for M410, M510, and M511 Printers
Étiquettes
White B-425A thermal transfer printable polypropylene is ideal for labelling applications requiring excellent solvent resistance and print performance.
  • Service temperature of −70 to +80 °C
  • Compatible with Brady printers BMP41, BMP51, BMP53, M410, M510, and M511
  • For use with ribbon series R4300
White B-425A thermal transfer printable polypropylene provides no visible affect to the label when exposed to most common chemicals, oils, or water.
Brady Authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability.
Certifications: UL approved; CSA accepted. Meets requirements of a halogen-free material per DIN VDE 0472 part 815.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité