Aggressive Adhesive Multi-Purpose Nylon Labels B-499 for M410, M510 and M511 Printers
Fournisseur: Brady
177011
BRDY177011EA 77.14 EUR InStock
BRDY177011
Aggressive Adhesive Multi-Purpose Nylon Labels B-499 for M410, M510 and M511 Printers
Étiquettes
Looking for a tough and versatile general-purpose label material? Look no further than the B-499 nylon cloth label material. Print almost anywhere with a Brady portable label printer.
  • Designed to withstand extreme temperatures, chemicals, greases, oils, cleaners and detergents
  • Permanent adhesive labels that stick to smooth, rough, and powder-coated surfaces
  • Compatible with Brady printers BMP41, BMP51, BMP53, M410, M510, and M511
  • For use with ribbon series R4300
Ideal for labelling tightly-curved surfaces such as wires and cables, as well as general-purpose lab, equipment panels, faceplates, and terminal blocks. Durable labels are designed to withstand extreme temperatures, chemicals, greases, oils, cleaners and detergents. The permanent adhesive sticks to smooth, rough, and powder-coated surfaces alike.
Certifications: UL Recognised to UL969, CSA to C22.2 No. 0.15-95, RoHS compliant to 2011/65/EU (Pass 2015/863).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité