Brady® StainerBondz™ Chemical-Resistant Polyester Slide Labels with 76 mm Core
Fournisseur: Brady
321703
BRDY321703EA 316.1 EUR InStock
BRDY321703
Brady® StainerBondz™ Chemical-Resistant Polyester Slide Labels with 76 mm Core
Étiquettes
Super chemical resistant polyester (B-488) is designed for use in slide staining processes and other laboratory identification requiring extreme chemical resistance.
  • B-488 performs well in commonly used H&E slide staining processes
  • Excellent resistance to various chemicals, oils and water
  • Operating temperature from ―80 to 130 °C
This B-488 material, combined with R-4300 ribbon, is the recipe for a solution that remains resistant to xylene and other chemicals commonly used in slide staining processes.
No print to wipe off, no buckling of the label, no discolouration that would make barcodes unreadable, and the label stays on the slide.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité