Magicard™ Pronto100 Printer
Fournisseur: Brady
321936
BRDY321936EA 700 EUR InStock
BRDY321936
Magicard™ Pronto100 Printer
Imprimantes Document Printers
Pronto100 printer delivers extreme performance with its high speed, optimum quality, and eco-friendly design and manufacturing.
  • High speed
  • High quality printing
  • Trusted identity
  • Comes with three standard templates
Pronto100 printer was developed to exceed the expectations of connected and tech-savvy customers. ​The Pronto100 printer offers the highest print speed of the most compact Magicard printers available today. With photorealistic colour reproduction and guaranteed edge-to-edge printing, your printed cards have never looked better. The input and output trays located at the front of the printer are easily accessible when the printer is on the receiving counter.
A HoloKote® security watermark can be printed over the entire surface of the card. Digital Shredding™ once data has been used for the print job, it is fragmented, making it unrecoverable and avoiding storage on the printer.
Information de livraison: This Pronto 100 printer comes with a power supply (with UK and EU cables), a USB cable, a cleaning card and a quick start guide. With MagiCover limited 3 years guarantee.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité